Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Chloro-4-nitrophenyl)homopiperazine, 96%, Thermo Scientific™

Catalog No. AAH6310503
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6310503 1 g
AAH63105MD 250 mg
AAH6310506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6310503 Supplier Thermo Scientific Chemicals Supplier No. H6310503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220220-19-9
Molecular Formula C11H14ClN3O2
Molecular Weight (g/mol) 255.702
InChI Key MVZOFBIWSFBCKA-UHFFFAOYSA-N
Synonym 1-2-chloro-4-nitrophenyl-1,4-diazepane, 1-2-chloro-4-nitro-phenyl-1,4 diazepane, 1h-1,4-diazepine, 1-2-chloro-4-nitrophenyl hexahydro, 1-2-chloro-4-nitrophenyl-1,4-diazaperhydroepine
PubChem CID 540495
IUPAC Name 1-(2-chloro-4-nitrophenyl)-1,4-diazepane
SMILES C1CNCCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

Specifications

Quantity 1 g
UN Number UN3259
Formula Weight 255.7
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)homopiperazine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.