Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Chloro-4-nitrophenyl)pyrrolidine, 97%, Thermo Scientific™

Catalog No. AAH63415MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63415MD 250 mg
AAH6341503 1 g
AAH6341506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63415MD Supplier Thermo Scientific Chemicals Supplier No. H63415MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55403-91-3
Molecular Formula C10H11ClN2O2
Molecular Weight (g/mol) 226.66
InChI Key XMLGZIMCJDYHOD-UHFFFAOYSA-N
Synonym 1-2-chloro-4-nitrophenyl pyrrolidine, 4-pyrrolidino-3-chloronitrobenzene, pyrrolidine, 1-2-chloro-4-nitrophenyl, 2-chloro-4-nitrophenyl pyrrolidine, cambridge id 5139302
PubChem CID 108707
IUPAC Name 1-(2-chloro-4-nitrophenyl)pyrrolidine
SMILES C1CCN(C1)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

Specifications

Quantity 250 mg
Formula Weight 226.66
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(2-Chloro-4-nitrophenyl)pyrrolidine
EINECSNumber 259-625-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.