Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Cyclohexanedione, 98%

Catalog No. p-4509425
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111160100 10 g Glass bottle
AC111160500 50 g Glass bottle
AC111162500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111160100 Supplier Thermo Scientific Chemicals Supplier No. 111160100
Only null left
Add to Cart

CAS: 765-87-7 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-87-7
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
InChI Key OILAIQUEIWYQPH-UHFFFAOYSA-N
Synonym 1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol
PubChem CID 13006
ChEBI CHEBI:41674
IUPAC Name cyclohexane-1,2-dione
SMILES C1CCC(=O)C(=O)C1

Specifications

Melting Point 35.0°C to 38.0°C
Color Yellow
Boiling Point 193.0°C to 195.0°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H8(=O)2
Refractive Index 1.5180 to 1.5200
Quantity 10 g
Beilstein 07,II,526
Fieser 02,93
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and diethyl ether
Formula Weight 112.13
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 1, 2-Cyclohexanedione
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 212-155-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.