Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diaminocyclohexane, mixture of isomers, 99%

Catalog No. p-7027695
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2465730 250 g
AAB246570B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2465730 Supplier Thermo Scientific Chemicals Supplier No. B2465730
Only null left
Add to Cart

CAS: 694-83-7 | C6H14N2 | 114.192 g/mol

1,2-Diaminocyclohexane was used in the synthesis of chiral ruthenium(IV)-oxo complexes & bicyclic pyrazine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diaminocyclohexane was used in the synthesis of chiral ruthenium(IV)-oxo complexes & bicyclic pyrazine derivatives.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, air, carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-83-7
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00001491
InChI Key SSJXIUAHEKJCMH-UHFFFAOYSA-N
Synonym 1,2-cyclohexanediamine, 1,2-diaminocyclohexane, 1,2-cylohexanediamine, cyclohex-1,2-ylenediamine, 1r,2r---1,2-diamino cyclohexane, dsstox_cid_7301, dsstox_rid_78396, dsstox_gsid_27301, +/--trans-1,2-diaminocyclohexane, cis-1,2-cyclohexandiamine
PubChem CID 4610
IUPAC Name cyclohexane-1,2-diamine
SMILES C1CCC(C(C1)N)N

Specifications

Density 0.94
pH 12
Boiling Point 191°C
Flash Point 75°C (167°F)
Odor Amine-like
Refractive Index 1.487
Quantity 250 g
UN Number UN2735
Beilstein 506142
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 1,2-Diaminocyclohexane, mixture of isomers
Show More Show Less
Hazard Category H227-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H317-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 211-776-7
RTECSNumber GU8749500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.