Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Dibromoethane

Catalog No. 501861143
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-186-1143 1 L
50-186-1142 100 mL
50-186-1144 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-186-1143 Supplier Sigma Aldrich Supplier No. 035051L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-Dibromoethane
TRUSTED_SUSTAINABILITY

Specifications

CAS 106-93-4
Melting Point 8°C to 11°C (lit.)
Density 2.18 g/mL (at 25°C (literature))
Boiling Point 131°C to 132°C (lit.)
Molecular Formula C2H4Br2
Refractive Index n20/D 1.539 (literature); n20/D 1.539
Linear Formula BrCH2CH2Br
MDL Number MFCD00000233
Quantity 1 L
Synonym EDB; Ethylene dibromide
Molecular Weight (g/mol) 187.86
Percent Purity ≥98.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.