Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloro-4-fluoro-5-nitrobenzene 98.0+%, TCI America™
SDP

Catalog No. D469425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D469425G 25 g
D46945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D469425G Supplier TCI America Supplier No. D469425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2339-78-8
Molecular Formula C6H2Cl2FNO2
Molecular Weight (g/mol) 209.985
MDL Number MFCD00075330
InChI Key FXOCDIKCKFOUDE-UHFFFAOYSA-N
Synonym 4,5-dichloro-2-fluoronitrobenzene, benzene, 1,2-dichloro-4-fluoro-5-nitro, buttpark 89\07-58, 4,5-dichloro-2-fluoro-1-nitrobenzene, pubchem4385, intermediates-zcf02626, ksc201k5p, 1,2-ichloro-4-luoro-5-itro-benzene
PubChem CID 533951
IUPAC Name 1,2-dichloro-4-fluoro-5-nitrobenzene
SMILES C1=C(C(=CC(=C1Cl)Cl)F)[N+](=O)[O-]

Specifications

Melting Point 17°C
Boiling Point 247°C
Quantity 25 g
Formula Weight 209.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dichloro-4-fluoro-5-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.