Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichlorobenzene-d{4}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog No. AA4228006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4228006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4228006 Supplier Thermo Scientific Chemicals Supplier No. 4228006
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2199-69-1 | C6H4Cl2 | 151.02 g/mol

It is applied in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2199-69-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 151.02
MDL Number MFCD00037106
InChI Key RFFLAFLAYFXFSW-RHQRLBAQSA-N
Synonym 1,2-dichlorobenzene-d4, 1,2-dichlorobenzene d4, benzene-1,2,3,4-d4-, 5,6-dichloro, o-dichloro 2h4 benzene, deuterated 1,2-dichlorobenzene, tetradeutero-1,2-dichlorobenzene, 1,2-dichlorobenzene-d4, 98 atom % d, 1,2-dichlorobenzene-d4,98atom%d, benzene-1,2,3,4-d4, 5,6-dichloro, 1,2-dichloro-3,4,5,6-tetradeuterobenzene
PubChem CID 519913
IUPAC Name 1,2-dichloro-3,4,5,6-tetradeuteriobenzene
SMILES [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]

Specifications

Physical Form Liquid
Quantity 5 g
Density 1.34 g/mL
Refractive Index 1.5509
Flash Point 65°C (150°F)
Melting Point -17°C
Boiling Point 181°C
UN Number UN1591
Solubility Information Insoluble in water
Formula Weight 151.03
Percent Purity 99%
Chemical Name or Material 1,2-Dichlorobenzene-d4, Isotopic
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: O-DICHLOROBENZENE
EINECSNumber 218-606-0
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.