CAS RN 2199-69-1
1,2-Dichlorobenzene-d4, for NMR, 99 atom % D
CAS: 2199-69-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 151.02 MDL Number: MFCD00037106 InChI Key: RFFLAFLAYFXFSW-RHQRLBAQSA-N IUPAC Name: dichloro(²H₄)benzene SMILES: [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]
1,2-Dichlorobenzene-d4 Solution, Certified Reference Material, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
1,2-Dichlorobenzene-d4, for NMR, 98 atom% D
CAS: 2199-69-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 151.02 MDL Number: MFCD00037106 InChI Key: RFFLAFLAYFXFSW-RHQRLBAQSA-N Synonym: 1,2-dichlorobenzene-d4,1,2-dichlorobenzene d4,benzene-1,2,3,4-d4-, 5,6-dichloro,o-dichloro 2h4 benzene,deuterated 1,2-dichlorobenzene,tetradeutero-1,2-dichlorobenzene,1,2-dichlorobenzene-d4, 98 atom % d,1,2-dichlorobenzene-d4,98atom%d,benzene-1,2,3,4-d4, 5,6-dichloro,1,2-dichloro-3,4,5,6-tetradeuterobenzene PubChem CID: 519913 IUPAC Name: 1,2-dichloro-3,4,5,6-tetradeuteriobenzene SMILES: [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]
1,2-Dichlorobenzene-d{4}, 99% (Isotopic), Thermo Scientific Chemicals
CAS: 2199-69-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 151.02 MDL Number: MFCD00037106 InChI Key: RFFLAFLAYFXFSW-RHQRLBAQSA-N Synonym: 1,2-dichlorobenzene-d4,1,2-dichlorobenzene d4,benzene-1,2,3,4-d4-, 5,6-dichloro,o-dichloro 2h4 benzene,deuterated 1,2-dichlorobenzene,tetradeutero-1,2-dichlorobenzene,1,2-dichlorobenzene-d4, 98 atom % d,1,2-dichlorobenzene-d4,98atom%d,benzene-1,2,3,4-d4, 5,6-dichloro,1,2-dichloro-3,4,5,6-tetradeuterobenzene PubChem CID: 519913 IUPAC Name: 1,2-dichloro-3,4,5,6-tetradeuteriobenzene SMILES: [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]
1,2-Dichlorobenzene-d4, SPEX CertiPrep™
CAS: 2199-69-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 151.02 MDL Number: MFCD00037106 InChI Key: RFFLAFLAYFXFSW-RHQRLBAQSA-N PubChem CID: 519913 IUPAC Name: 1,2-dichloro-3,4,5,6-tetradeuteriobenzene SMILES: [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]