Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichlorobenzene-d4, for NMR, 98 atom% D

Catalog No. AC321260050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC321260050 5 g Glass Bottle
AC321260010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC321260050 Supplier Thermo Scientific Chemicals Supplier No. 321260050
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

CAS: 2199-69-1 | C6H4Cl2 | 151.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2199-69-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 151.02
MDL Number MFCD00037106
InChI Key RFFLAFLAYFXFSW-RHQRLBAQSA-N
Synonym 1,2-dichlorobenzene-d4, 1,2-dichlorobenzene d4, benzene-1,2,3,4-d4-, 5,6-dichloro, o-dichloro 2h4 benzene, deuterated 1,2-dichlorobenzene, tetradeutero-1,2-dichlorobenzene, 1,2-dichlorobenzene-d4, 98 atom % d, 1,2-dichlorobenzene-d4,98atom%d, benzene-1,2,3,4-d4, 5,6-dichloro, 1,2-dichloro-3,4,5,6-tetradeuterobenzene
PubChem CID 519913
IUPAC Name 1,2-dichloro-3,4,5,6-tetradeuteriobenzene
SMILES [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]

Specifications

Physical Form Liquid
Color Colorless
Packaging Glass Bottle
Quantity 5 g
Water 0.2% max. (Coulometric)
Density 1.3400g/mL
Deuterium (D) 97.5% min.
Linear Formula C6D4Cl2
Formula Weight 151.04
Grade NMR
Chemical Name or Material 1, 2-Dichlorobenzene-d4, 98 atom% D
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.