Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichlorobenzene-d4, for NMR, 99 atom % D

Catalog No. AC464060010
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464060010 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC464060010 Supplier Thermo Scientific Chemicals Supplier No. 464060010
Add to Cart

CAS: 2199-69-1 | C6H4Cl2 | 151.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2199-69-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 151.02
MDL Number MFCD00037106
InChI Key RFFLAFLAYFXFSW-RHQRLBAQSA-N
IUPAC Name dichloro(²H₄)benzene
SMILES [2H]C1=C([2H])C([2H])=C(Cl)C(Cl)=C1[2H]

Specifications

Flash Point 65°C
Specific Gravity 1.34
Refractive Index 1.5509
Density 1.3400 g/mL
Quantity 1 mL
Linear Formula C6D4Cl2
Solubility Information Solubility aq. soln.: 0.13 g/L water (20°C). Other solubilities: miscible with most common organic solvents, miscible with alcohol, ether and benzene
Formula Weight 151.04
Grade NMR
Chemical Name or Material 1,2-Dichlorobenzene-d4
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Very toxic to aquatic life with long lasting effects.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

If eye irritation persists: Get medical advice/attention.

Avoid breathing dust/fume/gas/mist/vapors/spray.

Avoid release to the environment.

IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

EINECSNumber 218-606-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.