Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Difluorobenzene 98.0+%, TCI America™
SDP

Catalog No. D162725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D162725G 25 g
D16275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D162725G Supplier TCI America Supplier No. D162725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 367-11-3
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000284
InChI Key GOYDNIKZWGIXJT-UHFFFAOYSA-N
Synonym o-difluorobenzene, benzene, 1,2-difluoro, benzene, o-difluoro, ortho-difluorobenzene, o-difluoro benzene, difluorobenzene, 1,2-difluoro-benzene, unii-aw7qgmw29c, aw7qgmw29c, 0-difluorobenzene
PubChem CID 9706
ChEBI CHEBI:38583
IUPAC Name 1,2-difluorobenzene
SMILES FC1=CC=CC=C1F

Specifications

Color Yellow
Boiling Point 91°C
Quantity 25 g
UN Number 1993
Formula Weight 114.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Difluorobenzene
EINECSNumber (3)-2688
RTECSNumber CZ5655000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.