Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dihydronaphthalene, MilliporeSigma™ Supelco™

Catalog No. 111002100 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2100 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2100 Supplier MilliporeSigma Supelco Supplier No. 45778250MG

1,2-Dihydronaphthalene is a bicyclic hydrocarbon, which resembles naphthalene but shows partial unsaturation in one of its rings. Its derivatives find wide applications in natural compounds of therapeutic interest.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 447-53-0
Density 0.997 g/mL (at 25°C)
Flash Point 67°C
Physical Form Neat
Linear Formula C10H10
MDL Number MFCD00001672
Melting Point -8°C
Boiling Point 89°C (16 mmHg)
Type Analytical Standard
Quantity 250 mg
Formula Weight 130.19
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.