Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diiodobenzene (stabilized with Copper chip) 98.0+%, TCI America™
SDP

Catalog No. D060625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D060625G 25 g
D06065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D060625G Supplier TCI America Supplier No. D060625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 615-42-9
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.91
MDL Number MFCD00001038
InChI Key BBOLNFYSRZVALD-UHFFFAOYSA-N
Synonym o-diiodobenzene, benzene, 1,2-diiodo, benzene, o-diiodo, diiodobenzene, benzene, diiodo, benzene,2-diiodo, pubchem3831, 1,2-diiodobenzene
PubChem CID 11994
IUPAC Name 1,2-diiodobenzene
SMILES IC1=CC=CC=C1I

Specifications

Color Yellow
Boiling Point 152°C
Quantity 25 g
Formula Weight 329.91
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Diiodobenzene (stabilized with Copper chip)
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.