Learn More
Medchemexpress LLC 1,2-dilauroyl-sn-glycero-3-phosphoethanolamine | 59752-57-7 | MFCD00042828 | 98.0% | 579.75 | C29H58NO8P | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW32222525MG
Encompass_Preferred
1,2-Dilauroyl-sn-glycero-3-phosphoethanolamine (DLPE) is a phosphatidylethanolamine lipid bearing lauric acid (12:0) acyl chains. It is commonly used as a lipid component in the preparation of liposomes and other lipid-based formulations for research use only. The material is supplied as a white to off-white solid with high purity and specified storage conditions.
  • Phosphatidylethanolamine derivative with C12:0 acyl chains.
  • Suitable as a lipid component for liposome synthesis.
  • High purity for research applications.
  • Available in small laboratory quantities (e.g., 25 mg, 50 mg, 100 mg).
  • White to off-white solid; store powder at -20°C.
Catalog No. 50-002-82514
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.