Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethoxyethane, ≥99%, Reagent Grade, Inhibitor-free, Solstice

Catalog No. 60047985 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-985 20 L Stainless steel can
60-047-983 1 L Glass bottle
60-047-984 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-047-985 Supplier Solstice Supplier No. E2740820L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade, inhibitor-free,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-71-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00008502
InChI Key XTHFKEDIFFGKHM-UHFFFAOYSA-N
Synonym monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
PubChem CID 8071
ChEBI CHEBI:42263
IUPAC Name 1,2-dimethoxyethane
SMILES COCCOC

Specifications

Melting Point -58°C
Color Colorless
Density 0.867g/mL at 25°C
pH Neutral
Boiling Point 85°C (1013 hPa)
Flash Point −2°C (28.4°F)
Packaging Stainless steel can
Linear Formula CH3OCH2CH2OCH3
Quantity 20 L
UN Number UN2252
Beilstein 1209237
Vapor Density 3.1 (20°C, vs air)
Formula Weight 90.12g/mol
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material 1,2-Dimethoxyethane
Show More Show Less
Hazard Statement P201-P210-P280-P308 + P313-P370 + P378-P403 + P235
DOTInformation 3/PGII
RTECSNumber 203-794-9
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.