Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethoxyethane, CHROMASOLV™, for HPLC, 99.9%, inhibitor-free, Solstice

Catalog No. 60045846 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-846 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-846 Supplier Solstice Supplier No. 3074321L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glass-distilled, submicron-filtered solvent designed for HPLC instrumentation and organic synthesis applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-71-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00008502
InChI Key XTHFKEDIFFGKHM-UHFFFAOYSA-N
Synonym monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
PubChem CID 8071
ChEBI CHEBI:42263
IUPAC Name 1,2-dimethoxyethane
SMILES COCCOC

Specifications

CAS Max % ≤100.0000%
Melting Point -58°C (literature)
Density 0.867 g/mL at 25 °C(lit.)
Boiling Point 85°C
Flash Point −2°C
Packaging Glass Bottle
Linear Formula CH3OCH2CH2OCH3
Quantity 1 L
UN Number UN2252
Beilstein 1209237
Formula Weight 90.12
Percent Purity >99.85%
Grade HPLC
Physical Form Liquid
Chemical Name or Material 1,2-Dimethoxyethane, CHROMASOLV™, for HPLC, 99.9%, inhibitor-free
Show More Show Less
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: 1,2-Dimethoxyethane
EINECSNumber 203-794-9
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.