Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethylcyclohexane, Mixture of cis and trans 98%, Thermo Scientific™

Catalog No. AC116140010
Change view
Click to view available options
1L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC116140010 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC116140010 Supplier Thermo Scientific Chemicals Supplier No. 116140010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 583-57-3
CAS Min % 97.5
CAS Max % 100.0
Density 0.7780g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H16
Refractive Index 1.4270 to 1.4360 (20°C, 589nm)
Linear Formula C6H10(CH3)2
Quantity 1L
MDL Number MFCD00064172
Beilstein 05, 36
Synonym cyclohexane, 1,2-dimethyl, cyclohexane, 1,2-dimethyl-cis/trans, cyclohexane, 1,2-dimethyl-, 1r,2r-rel, hexahydro-o-xylene, o-dimethylcyclohexane, cyclohexane, 1,2-dimethyl-, 1r,2s-rel, cyclohexane,2-dimethyl, acmc-1an8x, acmc-209o4l, cyclohexane, 1,2-dimethyl-8ci 9ci
InChI Key KVZJLSYJROEPSQ-UHFFFAOYSA-N
SMILES CC1CCCCC1C
IUPAC Name 1,2-dimethylcyclohexane
Molecular Weight (g/mol) 112.216
PubChem CID 11416
Formula Weight 112.21
Percent Purity ≥97.5%
Chemical Name or Material 1, 2-Dimethylcyclohexane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-509-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.