Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 1,2-Dimyristoyl-sn-glycero-3-phosphorylglycerol Sodium Salt, Purity >98%, 100 mg

Catalog No. 502706055
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6055 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6055 Supplier Abcam Supplier No. AB143952100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity phosphatidylglycerol for use in lipid bilayer studies and biological systems.

  • Molecular Weight: 688.8 Da
  • Purity: >98%
  • Storage Conditions: Store at -20°C
  • Shipping Conditions: Ambient - Can Ship with Ice
  • Application: For use in lipid bilayer studies and biological systems

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% 1,2-Dimyristoyl-sn-glycero-3-phosphorylglycerol sodium salt
Content And Storage Store at -20°C.
Shipping Condition Ambient
Substrate Properties Lipid-based Substrate
Target Molecule Phosphatidylglycerol
Formulation 100% 1,2-Dimyristoyl-sn-glycero-3-phosphorylglycerol sodium salt
Quantity 100 mg
For Use With (Application) Lipid bilayer studies, biological systems
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.