Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Distearoyl-sn-glycero-3-phosphocholine 95.0+%, TCI America™
SDP

Catalog No. D39261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39261G 1 g
D3926200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39261G Supplier TCI America Supplier No. D39261G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 816-94-4
Molecular Formula C44H88NO8P
Molecular Weight (g/mol) 790.16
MDL Number MFCD00036905
InChI Key NRJAVPSFFCBXDT-HUESYALOSA-N
Synonym 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(stearoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, DSPC
PubChem CID 94190
ChEBI CHEBI:83718
IUPAC Name (2-{[(2R)-2,3-bis(octadecanoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
SMILES CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCCCC

Specifications

Melting Point 236°C
Color White
Quantity 1 g
Formula Weight 790.16
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Distearoyl-sn-glycero-3-phosphocholine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.