accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 816-94-4

H3COO(R)OPOOONH3CCH3H3COOCH3

CAS RN 816-94-4

IUPAC Name: (2-{[(2R)-2,3-bis(octadecanoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Synonyms: distearoyl phosphatidylcholine distearoyl glycerophosphocholine
Molecular Weight (g/mol): 790.16
Molecular Formula: C44H88NO8P
InChi Key: NRJAVPSFFCBXDT-HUESYALOSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)