Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxybutane, SPEX CertiPrep™
SDP

Catalog No. S1867 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S-1867 1 mL 2mL amber prescored glass ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S-1867 Supplier SPEX CertiPrep Supplier No. S1867

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2-Epoxybutane106-88-71000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 106-88-7 , 67-56-1
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
InChI Key RBACIKXCRWGCBB-UHFFFAOYSA-N
Synonym 1,2-epoxybutane, 1,2-butylene oxide, oxirane, ethyl, ethyloxirane, 1,2-butene oxide, ethylethylene oxide, 1-butene oxide, oxirane, 2-ethyl, butane, epoxy, 1-butylene oxide
PubChem CID 7834
ChEBI CHEBI:82326
IUPAC Name 2-ethyloxirane
SMILES CCC1CO1

Specifications

CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Packaging 2mL amber prescored glass ampule
Quantity 1 mL
Chemical Name or Material 1,2-Epoxybutane
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.