Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxycyclooctane 97.0+%, TCI America™
SDP

Catalog No. E05165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05165G 5 g
E051625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05165G Supplier TCI America Supplier No. E05165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 286-62-4
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00005164
InChI Key MELPJGOMEMRMPL-UHFFFAOYSA-N
Synonym 9-oxabicyclo 6.1.0 nonane, 1,2-epoxycyclooctane, cyclooctene oxide, epoxycyclooctane, cyclooctene, oxide, cyclooctane, 1,2-epoxy, ccris 3753, 9-oxabicyclo 6.1.0 nonane, cis, cyclooctane epoxide, cyclooctane,2-epoxy
PubChem CID 67513
IUPAC Name 9-oxabicyclo[6.1.0]nonane
SMILES C1CCCC2C(O2)CC1

Specifications

Melting Point 55°C
Color White
Boiling Point 75°C
Quantity 5 g
UN Number 1325
Formula Weight 126.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Epoxycyclooctane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.