Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Epoxycyclopentane 97.0+%, TCI America™
SDP

Catalog No. E051325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E051325G 25 g
E05135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E051325G Supplier TCI America Supplier No. E051325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 285-67-6
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00005161
InChI Key GJEZBVHHZQAEDB-UHFFFAOYNA-N
Synonym cyclopentene oxide, 6-oxabicyclo 3.1.0 hexane, 1,2-epoxycyclopentane, cyclopentane oxide, cyclopentene epoxide, epoxycyclopentane, cis-1,2-epoxycyclopentane, cyclopentane, 1,2-epoxy, 6-oxa-bicyclo 3.1.0 hexane
PubChem CID 9244
IUPAC Name 6-oxabicyclo[3.1.0]hexane
SMILES C1CC2OC2C1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 25 g
UN Number 1993
Formula Weight 84.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Epoxycyclopentane
RTECSNumber RN8935000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.