Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Methoxyphenoxy)-2-propanol 90.0+%, TCI America™
SDP

Catalog No. M106025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M106025G 25 g
M10605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M106025G Supplier TCI America Supplier No. M106025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64120-49-6
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00191539
InChI Key CUJJTCXPBXHBTJ-UHFFFAOYSA-N
Synonym 2-(2-Hydroxypropoxy)anisole
PubChem CID 22036327
IUPAC Name 1-(2-methoxyphenoxy)propan-2-ol
SMILES CC(COC1=CC=CC=C1OC)O

Specifications

Color Colorless
Quantity 25 g
Formula Weight 182.22
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Methoxyphenoxy)-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.