Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[(2-Methyl-4-thiazolyl)methyl]piperazine hydrochloride, Thermo Scientific™

Catalog No. AAH51021MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51021MD 250 mg
AAH5102103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51021MD Supplier Thermo Scientific Chemicals Supplier No. H51021MD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 880361-73-9
Molecular Formula C9H15N3S
Molecular Weight (g/mol) 197.30
MDL Number MFCD05864526,MFCD07771967
InChI Key RZWRPESYZFJDIU-UHFFFAOYSA-N
PubChem CID 16227460
IUPAC Name 2-methyl-4-(piperazin-1-ylmethyl)-1,3-thiazole
SMILES CC1=NC(CN2CCNCC2)=CS1

Specifications

Quantity 250 mg
Synonym 2-methyl-4-piperazin-1-ylmethyl thiazole, 1-2-methyl-1,3-thiazol-4-yl methyl piperazine, 1-2-methyl-thiazol-4-ylmethyl-piperazine, 1-2-methyl-thiazol-4-ylmethyl piperazine, 2-methyl-4-piperazin-1-ylmethyl-1,3-thiazole, 2-methyl-4-piperazinylmethyl-1,3-thiazole, 1-2-methylthiazol-4-ylmethyl piperazine, 1-2-methyl-4-thiazolyl methyl piperazine, piperazine,1-2-methyl-4-thiazolyl methyl, piperazine, 1-2-methyl-4-thiazolyl methyl
Formula Weight 233.76
Chemical Name or Material 1-[(2-Methyl-4-thiazolyl)methyl]piperazine hydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.