Learn More
Thomas Scientific 96 Well 1.1mL Deep Well Plate, Square Bottom
SDP
Supplier:  Thomas Scientific 1148F30
Encompass
Multi-Well / Deep-Well Plates. Ideal for sample collection, preparation, and long-term storage. HTS-friendly—compatible with most liquid-handling robots. Centrifugation to 4,000 x G. Manufactured from virgin polypropylene to ensure purity and sample integrity, chemical resistance, and compatibility with long-term storage needs. Alphanumeric indexing molded in on A-H and 1-12 axes. Optimum sample mixing and recovery with highly-polished wells and radiused corners. Raised rims on each well facilitate secure sealing with films and sealing mats. Bar coded plates available—please inquire. Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile plates are also pyrogen-free.
Catalog No. NC9572578