Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Naphthyl)ethanol 98.0+%, TCI America™
SDP

Catalog No. N078825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N078825G 25 g
N07885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N078825G Supplier TCI America Supplier No. N078825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7228-47-9
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00004111
InChI Key AXRKCRWZRKETCK-UHFFFAOYSA-N
Synonym 1-2-naphthyl ethanol, 1-naphthalen-2-yl ethanol, +/--1-2-naphthyl ethanol, 1-naphthalen-2-yl ethan-1-ol, alpha-methylnaphthalene-2-methanol, .alpha.-methyl-2-naphthalenemethanol, pubchem9427, acmc-209gxz, acmc-1azul, methyl 2-naphtylcarbinol
PubChem CID 98243
IUPAC Name 1-naphthalen-2-ylethanol
SMILES CC(C1=CC2=CC=CC=C2C=C1)O

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 170°C
Quantity 25 g
Formula Weight 172.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Naphthyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.