Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-O-Isopropylidene-alpha-d-glucofuranose | 18549-40-1 | 99.4% | 220.22 g/mol | C9H16O6 | 100 G
SDP

Supplier:  Medchemexpress LLC HYW087995100G

Encompass_Preferred

1,2-O-Isopropylidene-alpha-d-glucofuranose (mono-acetone glucose) is a protected carbohydrate derivative supplied as a white to off-white solid. It is used as an intermediate or reagent in organic synthesis and life-science research.

  • High purity (99.4%).
  • Solid, white to off-white appearance.
  • Available in 50 g, 100 g, 250 g, and 500 g pack sizes.
  • Soluble in DMSO and forms clear solutions.
  • Stable at room temperature for up to 3 years; stock solutions stable at -80°C or -20°C.

Catalog No. 50-002-08426


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.