Learn More
Medchemexpress LLC 1,2-O-isopropylidene-alpha-D-xylofuranose | 20031-21-4 | MFCD00063295 | 99.9% | 190.19 g/mol | C8H14O5 | 100 G
Supplier: Medchemexpress LLC HYW015179100G
1,2-O-Isopropylidene-alpha-D-xylofuranose is an isopropylidene-protected derivative of D-xylose used as a synthetic building block in carbohydrate chemistry and organic synthesis. It is supplied as a solid (white to off-white) at high purity for research use and is commonly employed as an intermediate in stereoselective and asymmetric reactions.
- Isopropylidene-protected sugar intermediate for carbohydrate synthesis.
- High purity suitable for research applications.
- Solid, white to off-white for easy handling and storage.
- Soluble in DMSO with ultrasonic assistance for solution preparations.
- Useful in stereoselective and asymmetric transformations.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.