Learn More
Medchemexpress LLC 1,2-O-isopropylidene-alpha-D-xylofuranose | 20031-21-4 | MFCD00063295 | 99.9% | 190.19 | C8H14O5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01517910G
Encompass_Preferred
1,2-O-Isopropylidene-alpha-D-xylofuranose is an acetonide-protected derivative of D-xylose supplied as a white to off-white solid for use in synthetic and carbohydrate chemistry. It serves as a chiral building block and protecting-group intermediate suitable for glycosylation and asymmetric synthesis, with reported use in the asymmetric alkylation of benzaldehyde with diethylzinc.
  • Acetonide-protected D-xylose derivative for synthesis.
  • White to off-white solid suitable for laboratory use.
  • High purity for reliable synthetic performance.
  • Useful as a chiral building block in carbohydrate chemistry.
  • Reported application in asymmetric alkylation reactions.
Catalog No. 50-003-02647
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.