Learn More
Medchemexpress LLC 1,2-O-isopropylidene-alpha-D-xylofuranose | 20031-21-4 | MFCD00063295 | 99.9% | 190.19 g/mol | C8H14O5 | 25 G

Supplier: Medchemexpress LLC 25GHYW015179
1,2-O-Isopropylidene-alpha-D-xylofuranose is a protected derivative of D-xylose used as a building block in organic synthesis. It is reported for use in asymmetric alkylation reactions and is supplied as a high-purity research reagent with supporting analytical documentation.
- Protected derivative of D-xylose for synthetic chemistry
- Reported for asymmetric alkylation of benzaldehyde with diethylzinc
- High purity: 99.93%
- Molecular weight: 190.19 g/mol
- Molecular formula: C8H14O5
- Available in multiple pack sizes from 5 g to 100 g and larger by quote
- Analytical documents available, including COA, H NMR, GC, and SDS
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.