Learn More
Medchemexpress LLC 1,2-O-isopropylidene-alpha-D-glucofuranose (mono-acetone glucose) | 18549-40-1 | MFCD00063244 | 99.4% | 220.22 g/mol | C9H16O6 | 50 G
Supplier: Medchemexpress LLC HYW08799550G
1,2-O-isopropylidene-alpha-D-glucofuranose, also called mono-acetone glucose, is a protected derivative of D-glucose used as a reagent and building block in carbohydrate chemistry and organic synthesis. It is supplied as a white to off-white solid for laboratory research and synthetic applications.
- Protected glucose derivative used as a building block in carbohydrate chemistry.
- Useful intermediate for synthesis and derivatization of carbohydrates.
- High listed purity of 99.4%.
- Molecular weight 220.22 g/mol and formula C9H16O6.
- Available in lab-scale quantities such as 50 G for research use.