Learn More
Medchemexpress LLC 1,2-O-isopropylidene-alpha-D-xylofuranose | 20031-21-4 | MFCD00063295 | 99.9% | 190.19 g·mol⁻¹ | C8H14O5 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01517950G
Encompass_Preferred
1,2-O-isopropylidene-alpha-D-xylofuranose is an isopropylidene-protected derivative of D-xylofuranose used as a building block and intermediate in organic synthesis, particularly in carbohydrate chemistry and asymmetric alkylation reactions.
  • Isopropylidene-protected sugar useful for selective functionalization.
  • Suitable intermediate for glycoside and oligosaccharide synthesis.
  • Compatible with asymmetric alkylation and other carbon-carbon bond-forming reactions.
  • High purity suitable for research applications.
  • Solid, white to off-white appearance for easy handling.
  • Available in laboratory-scale pack sizes.
Catalog No. 50-003-03664
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.