Learn More
Medchemexpress LLC 1,2-O-isopropylidene-alpha-D-xylofuranose | 20031-21-4 | MFCD00063295 | 99.9% | 190.19 g·mol⁻¹ | C8H14O5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0151795G
Encompass_Preferred
1,2-O-isopropylidene-alpha-D-xylofuranose is a protected derivative of D-xylose used as a building block in carbohydrate and organic synthesis. It functions as a protected sugar intermediate for stereoselective transformations and functional-group manipulations in multistep syntheses.
  • Protected sugar derivative suitable for organic synthesis.
  • Useful in stereoselective alkylation reactions, for example benzaldehyde alkylation.
  • High purity (99.9%).
  • Molecular formula C8H14O5; molecular weight 190.19 g·mol⁻¹.
  • White to off-white solid physical form.
  • Available in small laboratory pack sizes for research use.
Catalog No. 50-003-03206