Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Octanediol, 97%

Catalog No. AAL0803109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0803109 10 g
AAL0803118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0803109 Supplier Thermo Scientific Chemicals Supplier No. L0803109
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1117-86-8 | C8H18O2 | 146.23 g/mol

1,2-Octanediol was used as organic modifier to improve the HPLC separation of organic acids and bases. It was also used in preparation of halohydrin palmitates. It is useful for treating head louse infestation clinically. It is also used in coating materials, slurries, paper mills and water circulation systems for the effective preservation against bacteria and fungi. It is used as an emollient, humectant, and wetting agent in cosmetic and skin care products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (3 g/L) at 20°C.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1117-86-8
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00010738
InChI Key AEIJTFQOBWATKX-UHFFFAOYSA-N
Synonym 1,2-octanediol, 1,2-dihydroxyoctane, 1,2-octylene glycol, capyryl glycol, racemic 1,2-octanediol, acmc-2099as, dsstox_cid_16646, dsstox_rid_79289
PubChem CID 14231
ChEBI CHEBI:34056
IUPAC Name octane-1,2-diol
SMILES CCCCCCC(CO)O

Specifications

Melting Point 28°C to 34°C
Boiling Point 131°C to 133°C (10 mmHg)
Flash Point 138°C (280°F)
Quantity 10 g
Beilstein 1719619
Solubility Information Soluble in water (3g/L) at 20°C.
Formula Weight 146.23
Percent Purity 97%
Chemical Name or Material 1,2-Octanediol
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 214-254-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.