Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 1-(2-Pyridylazo)-2-naphthol, 98%, pure, indicator grade

Catalog No. AC146310100
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC146310100 10 g Glass bottle
AC146310050 5 g Glass bottle
AC146310500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC146310100 Supplier Thermo Scientific Chemicals Supplier No. 146310100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 85-85-8 | C15H11N3O | 249.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-85-8
InChI Key RAXUMGMWXZYADR-OBGWFSINSA-N
Synonym 1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl
PubChem CID 5376264
IUPAC Name (1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3

Specifications

Melting Point 138.0°C to 141.0°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C15H11N3O
Quantity 10 g
Beilstein 22,I,694
Merck Index 15,7107
Solubility Information Solubility in water: insoluble.
Molecular Weight (g/mol) 249.27
Formula Weight 249.27
Percent Purity 98%
Grade Pure
Physical Form Powder
Chemical Name or Material 1-(2-Pyridylazo)-2-naphthol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 201-637-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.