Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Pyrimidyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. P111025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P111025G 25 g
P11105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P111025G Supplier TCI America Supplier No. P111025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20980-22-7
Molecular Formula C8H12N4
Molecular Weight (g/mol) 164.212
MDL Number MFCD00040742
InChI Key MRBFGEHILMYPTF-UHFFFAOYSA-N
Synonym 2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine
PubChem CID 88747
IUPAC Name 2-piperazin-1-ylpyrimidine
SMILES C1CN(CCN1)C2=NC=CC=N2

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 164.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-(2-Pyrimidyl)piperazine
RTECSNumber UV9702000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.