Learn More
Cayman Chemical 1 2DIarachIdoylracglycRol 10mg
SDP
Supplier:  Cayman Chemical 2693210 MG
Encompass_Preferred
A CB2 agonist (Ki 40 3 nM) selectively activates CB2 over CB1 in a GTPYS binding assay (EC50s 20 1 and 100000 nM respectively) reduces differences in hind paw weight bearing between MIA- and saline-injected knees in a rat model of knee joint-related chronic pain induced by MIA at 0 1 mg/kg dose-dependently reduces paclitaxel-induced mechanical and cold allodynia in mice and prevents development of morphine tolerance at 3 mg/kg per day for 12 days reduces paclitaxel-induced allodynia in morphine-tolerant mice
Catalog No. 50-293-7638
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.