Learn More
Cayman Chemical 1 2DIoloyl3Docosohexaenoy 25mg
SDP
Supplier:  Cayman Chemical 2682425 MG
Encompass_Preferred
A DYRK1A inhibitor (Ki 180 nM IC50 240 nM) selective for DYRK1A over MAOA and B (100 M) inhibits DYRK1A phosphorylation of tau in COS-7 cells expressing EGFP-DYRK1A and EGFP-tau in a concentration-dependent manner induces proliferation of human and ratBcells and increases insulin secretion by human islets in vitro reverses developmental deformities induced by DYRK1A overexpression in X. laevis embryos
Catalog No. 50-293-2696
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.