Learn More
Cytiva Fluorodyne EX EDF Membrane in Mini Kleenpak™ Capsules. 1/2IN-3/4IN
Supplier:  Cytiva KA02EDFP8G
Encompass
For smaller scale production Fluorodyne EX EDF Membrane in Mini Kleenpak™ Capsules maintains the high efficiency standard of other filters in larger scale range. Compact with asymmetric construction allows the capsule to minimize hold up volume and increase protein transmission. Complying with cGMP regulation the capsules are integrity tested. The products are the smallest components in Pall's UpScale Program line.
Catalog No. NC2091408
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.