Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,3-Trimethyl-2-methyleneindoline 96.0+%, TCI America™
SDP

Catalog No. T0879100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0879100ML 100 mL
T087925ML 25 mL
T0879500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0879100ML Supplier TCI America Supplier No. T0879100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 118-12-7
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.259
MDL Number MFCD00005813
InChI Key ZTUKGBOUHWYFGC-UHFFFAOYSA-N
Synonym 1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene
PubChem CID 8351
IUPAC Name 1,3,3-trimethyl-2-methylideneindole
SMILES CC1(C(=C)N(C2=CC=CC=C21)C)C

Specifications

Melting Point -10°C
Color Red
Boiling Point 248°C
Quantity 100 mL
UN Number 3082
Formula Weight 173.26
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,3-Trimethyl-2-methyleneindoline
RTECSNumber NM1926500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.