Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone 97.0+%, TCI America™
SDP

Catalog No. D50641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50641G 1 g
D50645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50641G Supplier TCI America Supplier No. D50641G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 277299-70-4
Molecular Formula C12H14N2O
Molecular Weight (g/mol) 202.257
MDL Number MFCD09795980
InChI Key HBWBJCSXUJIDGN-UHFFFAOYSA-N
PubChem CID 11735829
IUPAC Name 2-(3,4-dimethylphenyl)-5-methyl-1H-pyrazol-3-one
SMILES CC1=C(C=C(C=C1)N2C(=O)C=C(N2)C)C

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Powder at 20°C
Formula Weight 202.26
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.