Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5,5-Tetramethyl-1,3-cyclohexadiene 92.0+%, TCI America™
SDP

Catalog No. T17181ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17181ML 1 mL
T17185ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17181ML Supplier TCI America Supplier No. T17181ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4724-89-4
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00045518
InChI Key SZHAWDAGEJWQJK-UHFFFAOYSA-N
PubChem CID 78453
IUPAC Name 1,3,5,5-tetramethylcyclohexa-1,3-diene
SMILES CC1=CC(=CC(C1)(C)C)C

Specifications

Color Yellow
Quantity 1 mL
UN Number 3295
Synonym 1,3,5,5-tetramethyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1,3,5,5-tetramethyl, acmc-1alu2, 1, 1,3,5,5-tetramethyl, 1,3,5,5-tetramethylcyclohexadiene, 1,5,5-tetramethyl-1,3-cyclohexadiene, 1,3-cyclohexadiene,1,3,5,5-tetramethyl
Formula Weight 136.24
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5,5-Tetramethyl-1,3-cyclohexadiene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.