Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Tri(9H-carbazol-9-yl)benzene (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. T19345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19345G 5 g
T19341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19345G Supplier TCI America Supplier No. T19345G

Chemical Identifiers

CAS 148044-07-9
Molecular Formula C42H27N3
Molecular Weight (g/mol) 573.70
MDL Number MFCD03844810
InChI Key DVNOWTJCOPZGQA-UHFFFAOYSA-N
PubChem CID 22218951
IUPAC Name 9-[3,5-bis(9H-carbazol-9-yl)phenyl]-9H-carbazole
SMILES C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC(=CC(=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 330°C
Color White
Quantity 5 g
Formula Weight 573.70
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tri(9H-carbazol-9-yl)benzene (purified by sublimation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.