Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Trifluorobenzene 98.0+%, TCI America™
SDP

Catalog No. T103925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T103925G 25 g
T10395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T103925G Supplier TCI America Supplier No. T103925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 372-38-3
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.085
MDL Number MFCD00000333
InChI Key JXUKFFRPLNTYIV-UHFFFAOYSA-N
Synonym benzene, 1,3,5-trifluoro, sym-trifluorobenzene, unii-bn94c411f8, pubchem1069, 1,5-trifluorobenzene, acmc-209iru, 2,4,6-trifluorophenyl, benzene,3,5-trifluoro, 1,3,5-tri-fluorobenzene, 1,3,5-trifluoro-benzene
PubChem CID 9745
IUPAC Name 1,3,5-trifluorobenzene
SMILES C1=C(C=C(C=C1F)F)F

Specifications

Color Colorless
Boiling Point 76°C
Quantity 25 g
UN Number 1993
Formula Weight 132.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Trifluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.