Learn More
Medchemexpress LLC 1,3,5-trimethoxybenzene | 621-23-8 | MFCD00008385 | 168.19 g/mol | C9H12O3 | 25g
SDP
Supplier:  Medchemexpress LLC HYY067825G
Encompass_Preferred
1 3 5-Trimethoxybenzene (TRIMETHYL PHLOROGLUCINOL) is an electrophilic substitution reaction substrate targeting free chlorine (Cl ) and free bromine (Br ) 1 3 5-Trimethoxybenzene has highly selective electrophilic addition characteristics By capturing halogens it undergoes specific substitution reactions to generate stable halogenated products 1 3 5-Trimethoxybenzene can not only quench residual oxidants but also quantify the halogen concentration by detecting the product without affecting the stability of redox-sensitive disinfection byproducts (DBPs) 1 3 5-Trimethoxybenzene is mainly used in water quality testing and quantitative analysis of free chlorine/bromine in water At the same time in phytochemistry it is a key component of rose fragrance and participates in the study of pollination attraction mechanism[1][2]
Catalog No. 50-002-69883