Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Trimethoxybenzene, 99%

Catalog No. ac160550100
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160550100 10 g Glass bottle
AC160550500 50 g Glass bottle
AC160552500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC160550100 Supplier Thermo Scientific Chemicals Supplier No. 160550100
Only null left
Add to Cart

CAS: 621-23-8 | C9H12O3 | 168.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-23-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00008385
InChI Key LKUDPHPHKOZXCD-UHFFFAOYSA-N
Synonym phloroglucinol trimethyl ether, sym-trimethoxybenzene, benzene, 1,3,5-trimethoxy, 1,3,5-trimethyoxybenzene, unii-00vji3vg3d, 1,3,5-trimethoxy benzene, o,o,o-1,3,5-trimethylresorcinol, 00vji3vg3d, dsstox_cid_25963, dsstox_rid_81259
PubChem CID 69301
ChEBI CHEBI:31038
IUPAC Name 1,3,5-trimethoxybenzene
SMILES COC1=CC(OC)=CC(OC)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50.0°C to 53.0°C
Color White to Cream
Density 85g/mL
Boiling Point 255.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H3(OCH3)3
Quantity 10 g
Beilstein 06, 1101
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents,soluble in methanol
Formula Weight 168.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3, 5-Trimethoxybenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 210-673-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.