Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Trimethylpyrazole 98.0+%, TCI America™
SDP

Catalog No. T272425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T272425G 25 g
T27245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T272425G Supplier TCI America Supplier No. T272425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1072-91-9
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00015536
InChI Key HNOQAFMOBRWDKQ-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazole, 1h-pyrazole, 1,3,5-trimethyl, pyrazole, 1,3,5-trimethyl, akos pao-1133, pubchem20856, vitas-bb tbb000655, buttpark 15357-40, ksc490o8l, 1,3,5-trimethylpyrazole, 1,3,5-trimethyl-1h-pyrazole #
PubChem CID 14081
IUPAC Name 1,3,5-trimethylpyrazole
SMILES CC1=CC(=NN1C)C

Specifications

Melting Point 37°C
Color Yellow
Boiling Point 170°C
Quantity 25 g
UN Number 1325
Formula Weight 110.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,3,5-Trimethylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.