Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Undecatriene (mixture of isomers) 75.0+%, TCI America™
SDP

Catalog No. U006225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U006225ML 25 mL
U00625ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U006225ML Supplier TCI America Supplier No. U006225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16356-11-9
Molecular Formula C11H18
Molecular Weight (g/mol) 150.265
MDL Number MFCD00015699
InChI Key JQQDKNVOSLONRS-JEGFTUTRSA-N
PubChem CID 5367412
IUPAC Name (3E,5E)-undeca-1,3,5-triene
SMILES CCCCCC=CC=CC=C

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 mL
Formula Weight 150.27
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Undecatriene (mixture of isomers)
RTECSNumber YQ2833000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.