Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,6,8-Tetraphenylpyrene 98.0+%, TCI America™
SDP

Catalog No. T3042200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3042200MG 200 mg
T304250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3042200MG Supplier TCI America Supplier No. T3042200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13638-82-9
Molecular Formula C40H26
Molecular Weight (g/mol) 506.648
MDL Number MFCD00021662
InChI Key SIJHJHYRYHIWFW-UHFFFAOYSA-N
PubChem CID 256832
IUPAC Name 1,3,6,8-tetraphenylpyrene
SMILES C1=CC=C(C=C1)C2=CC(=C3C=CC4=C(C=C(C5=C4C3=C2C=C5)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8

Specifications

Melting Point 299°C
Color Yellow
Quantity 200 mg
Formula Weight 506.65
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,6,8-Tetraphenylpyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.